siRNA Details
Gene Symbol: LINC01115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033880.3 | 3, 4 | 777 | |||
NR_111963.1 | 2, 3 | 540 | ||||
GenBank mRNA | BC043553.1 | - | 3, 4 | 778 | ||
BC128532.1 | - | 2, 3 | 305 |
