siRNA Details
Gene Symbol: LINC01021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038848.1 | 2 | 300 | |||
GenBank mRNA | BC046201.1 | - | 2 | 300 | ||
BC101438.1 | - | 2 | 285 | |||
BC101982.1 | - | 2 | 285 | |||
CQ819099.1 | - | 2 | 308 | |||
CQ819101.1 | - | 2 | 308 |
