siRNA Details
Gene Symbol: C12orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080406.1 | NP_001073875.1 | 1 | 252 | ||
XM_011520847.2 | XP_011519149.1 | 2 | 794 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C12orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080406.1 | NP_001073875.1 | 1 | 252 | ||
XM_011520847.2 | XP_011519149.1 | 2 | 794 |