siRNA Details
Gene Symbol: THNSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018271.4 | NP_060741.3 | 6, 7 | 1082 | ||
XM_005264400.4 | XP_005264457.1 | 7, 8 | 1194 | |||
XM_005264401.4 | XP_005264458.1 | 7, 8 | 1163 | |||
XM_005264402.3 | XP_005264459.1 | 6, 7 | 2983 | |||
XM_006712041.3 | XP_006712104.1 | 6, 7 | 2983 | |||
XM_006712042.3 | XP_006712105.1 | 6, 7 | 2983 | |||
XM_011532953.2 | XP_011531255.1 | 6, 7 | 2983 | |||
XM_011532954.2 | XP_011531256.1 | 5, 6 | 2788 | |||
XM_011532955.2 | XP_011531257.1 | 6, 7 | 928 | |||
GenBank mRNA | AK001778.1 | - | 6, 7 | 702 | ||
AK095303.1 | - | 6, 7 | 2762 | |||
BC035315.1 | - | 6, 7 | 913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
