siRNA Details
Gene Symbol: ZNF879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136116.1 | NP_001129588.1 | 5 | 2111 | ||
XM_005265908.3 | XP_005265965.1 | 5 | 2805 | |||
XM_011534550.2 | XP_011532852.1 | 5 | 2140 | |||
XM_011534551.2 | XP_011532853.1 | 5 | 2256 | |||
GenBank mRNA | AK300504.1 | - | 5 | 2043 | ||
BX114675.1 | - | 1 | 27 | |||
BX648737.1 | - | 5 | 2081 |
