siRNA Details
Gene Symbol: LOC100507205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038309.1 | 1 | 31 | |||
GenBank mRNA | DB461660.1 | - | 1 | 31 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100507205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038309.1 | 1 | 31 | |||
GenBank mRNA | DB461660.1 | - | 1 | 31 |