siRNA Details
Gene Symbol: TMEM233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136534.1 | NP_001130006.1 | 1 | 49 | ||
XM_005253880.3 | XP_005253937.1 | 1 | 1385 | |||
XM_005253881.3 | XP_005253938.1 | 1 | 1381 | |||
XM_011538328.2 | XP_011536630.1 | 1 | 1382 | |||
XM_011538329.2 | XP_011536631.1 | 1 | 1382 | |||
XM_011538330.2 | XP_011536632.1 | 1 | 1382 | |||
XM_011538331.2 | XP_011536633.1 | 1 | 1380 |
