siRNA Details
Gene Symbol: SHTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127211.2 | NP_001120683.1 | 16 | 2151 | ||
NM_001258298.1 | NP_001245227.1 | 15 | 2098 | |||
NM_001258299.1 | NP_001245228.1 | 16 | 2151 | |||
NM_001258300.1 | NP_001245229.1 | 17 | 1941 | |||
XM_005270002.3 | XP_005270059.1 | 16 | 1969 | |||
XM_006717931.3 | XP_006717994.1 | 16 | 1969 | |||
XM_017016462.1 | XP_016871951.1 | 16 | 1661 | |||
GenBank mRNA | AK001984.1 | - | 2 | 245 | ||
AK091381.1 | - | 6 | 784 | |||
AK091578.1 | - | 15 | 1857 | |||
AK095419.1 | - | 6 | 610 | |||
AK124745.1 | - | 1 | 1810 | |||
AK302705.1 | - | 17 | 1941 | |||
AK304865.1 | - | 15 | 1745 | |||
BC022348.1 | - | 5 | 493 | |||
BC032303.1 | - | 16 | 1709 | 5169981 | ||
CR749417.1 | - | 16 | 1759 | |||
CX783645.1 | - | 7 | 722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
