siRNA Details
Gene Symbol: CFAP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164496.1 | NP_001157968.1 | 27 | 4338 | ||
XM_005247617.3 | XP_005247674.1 | 10 | 2615 | |||
XM_006713696.2 | XP_006713759.1 | 28 | 4928 | |||
XM_011512977.2 | XP_011511279.1 | 26 | 4556 | |||
XM_011512978.2 | XP_011511280.1 | 23 | 3825 | |||
XM_011512979.2 | XP_011511281.1 | 22 | 3569 | |||
XM_017006842.1 | XP_016862331.1 | 28 | 4928 | |||
XM_017006843.1 | XP_016862332.1 | 28 | 4928 | |||
XM_017006844.1 | XP_016862333.1 | 27 | 4772 | |||
XM_017006845.1 | XP_016862334.1 | 23 | 4046 | |||
XM_017006846.1 | XP_016862335.1 | 22 | 4723 | |||
XM_017006847.1 | XP_016862336.1 | 23 | 3766 | |||
GenBank mRNA | AK002004.1 | - | 1 | 133 | ||
AL133606.1 | - | 9 | 1680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
