siRNA Details
Gene Symbol: KIF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194313.2 | NP_919289.2 | 2 | 582 | ||
XM_011517861.1 | XP_011516163.1 | 3 | 803 | |||
XM_011517863.2 | XP_011516165.1 | 2 | 1892 | |||
XM_017014697.1 | XP_016870186.1 | 2 | 896 | |||
XM_017014698.1 | XP_016870187.1 | 2 | 767 | |||
GenBank mRNA | AK310014.1 | - | 3 | 803 | ||
BU660279.1 | - | 2 | 586 | |||
DT932659.1 | - | 1 | 126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
