siRNA Details
Gene Symbol: C2orf81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145054.1 | NP_001138526.1 | 4 | 1142 | ||
NM_001316764.1 | NP_001303693.1 | 3 | 984 | |||
NM_001316765.1 | NP_001303694.1 | 3 | 837 | |||
NM_001316766.1 | NP_001303695.1 | 2 | 1067 |
