siRNA Details
Gene Symbol: ZNF385D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534122.1 | XP_011532424.1 | 2 | 232 | ||
XM_011534123.2 | XP_011532425.1 | 2 | 232 | |||
XM_011534124.2 | XP_011532426.1 | 2 | 232 | |||
XM_017007191.1 | XP_016862680.1 | 2 | 232 | |||
XM_017007192.1 | XP_016862681.1 | 2 | 232 | |||
XM_017007194.1 | XP_016862683.1 | 2 | 223 | |||
XM_017007203.1 | XP_016862692.1 | 2 | 232 | |||
GenBank mRNA | AK310298.1 | - | 2 | 223 |
