siRNA Details
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001481.2 | NP_001001481.2 | 4, 5 | 339 | ||
NM_001271015.1 | NP_001257944.1 | 3, 4 | 306 | |||
NM_018299.4 | NP_060769.4 | 3, 4 | 306 | |||
GenBank mRNA | AK001873.1 | - | 3, 4 | 221 | ||
AK021735.1 | - | 3, 4 | 221 | |||
AK024050.1 | - | 4, 5 | 304 | |||
AK225554.1 | - | 3, 4 | 237 | |||
AK295792.1 | - | 3, 4 | 306 | |||
AY948289.1 | - | 4, 5 | 275 | |||
BC010900.1 | - | 3, 4 | 225 | 4081461 | ||
BC016326.1 | - | 3, 4 | 260 | 4093131 | ||
CR457275.1 | - | 2, 3 | 218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
