siRNA Details
Gene Symbol: ANKUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144960.2 | NP_001138432.1 | 5 | 1043 | ||
NM_001315505.1 | NP_001302434.1 | 5 | 933 | |||
NM_001315506.1 | NP_001302435.1 | 5 | 1043 | |||
XM_011512798.1 | XP_011511100.1 | 5 | 540 | |||
XM_011512799.1 | XP_011511101.1 | 5 | 774 | |||
XM_011512801.1 | XP_011511103.1 | 3 | 211 | |||
GenBank mRNA | AK027233.1 | - | 2 | 42 | ||
AK304359.1 | - | 5 | 774 | |||
BC171805.1 | - | 5 | 933 |
