siRNA Details
Gene Symbol: RNF121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300926.1 | NP_001287855.1 | 3, 4 | 708 | ||
NM_018320.4 | NP_060790.2 | 4, 5 | 746 | |||
NR_024147.1 | 3, 4 | 708 | ||||
NR_024148.1 | 4, 5 | 846 | ||||
XM_006718630.1 | XP_006718693.1 | 5, 6 | 559 | |||
XM_011545150.1 | XP_011543452.1 | 5, 6 | 591 | |||
GenBank mRNA | AK023139.1 | - | 4, 5 | 412 | ||
AK094508.1 | - | 4, 5 | 512 | |||
AL833638.1 | - | 3, 4 | 383 | |||
BC009672.2 | - | 4, 5 | 414 | |||
BC093084.1 | - | 3, 4 | 348 | |||
BC117273.1 | - | 4, 5 | 378 | |||
BG428563.1 | - | 4, 5 | 434 | |||
CR457298.1 | - | 4, 5 | 406 | |||
DA694800.1 | - | 5, 6 | 560 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
