siRNA Details
Gene Symbol: CCDC192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317938.1 | NP_001304867.1 | 7 | 861 | ||
XM_017009805.1 | XP_016865294.1 | 6 | 923 | |||
XM_017009806.1 | XP_016865295.1 | 7 | 827 | |||
XM_017009807.1 | XP_016865296.1 | 7 | 818 | |||
XM_017009808.1 | XP_016865297.1 | 6 | 749 | |||
XM_017009812.1 | XP_016865301.1 | 6 | 768 |
