siRNA Details
Gene Symbol: LOC100507387 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038402.1 | 3 | 453 | |||
GenBank mRNA | AK055006.1 | - | 1 | 72 | ||
AL117630.1 | - | 3 | 451 | |||
DA164303.1 | - | 3 | 513 |
Gene Symbol: FAM153A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB018295.1 | - | 24 | 2138 |
