siRNA Details
Gene Symbol: SHTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127211.2 | NP_001120683.1 | 16, 17 | 2168 | ||
NM_001258298.1 | NP_001245227.1 | 15, 16 | 2115 | |||
XM_017016462.1 | XP_016871951.1 | 16, 17 | 1678 | |||
GenBank mRNA | AK001984.1 | - | 2, 3 | 262 | ||
AK091381.1 | - | 6, 7 | 801 | |||
AK091578.1 | - | 15, 16 | 1874 | |||
AK095419.1 | - | 6, 7 | 627 | |||
AK304865.1 | - | 15, 16 | 1762 | |||
BC022348.1 | - | 5, 6 | 510 | |||
CR749417.1 | - | 16, 17 | 1776 | |||
CX783645.1 | - | 7, 8 | 739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
