siRNA Details
Gene Symbol: DDX19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320522.1 | NP_001307451.1 | 6 | 610 | ||
NM_001320525.1 | NP_001307454.1 | 8 | 764 | |||
NM_001320526.1 | NP_001307455.1 | 7 | 829 | |||
NM_001320527.1 | NP_001307456.1 | 6 | 567 | |||
NM_018332.4 | NP_060802.1 | 7 | 703 | |||
GenBank mRNA | AB209401.1 | - | 1 | 1771 | ||
AK001988.1 | - | 7 | 661 | |||
AK223262.1 | - | 7 | 627 | |||
AK299504.1 | - | 4 | 293 | |||
AK299539.1 | - | 6 | 495 | |||
AK310604.1 | - | 6 | 491 | |||
AK310808.1 | - | 2 | 405 | |||
AL832901.1 | - | 3 | 187 | |||
AL832970.1 | - | 7 | 614 | |||
BC005162.2 | - | 7 | 611 | 3530064 | ||
BC006544.2 | - | 7 | 611 | 2964377 | ||
BC137496.1 | - | 7 | 589 | 9021496 | ||
BC137497.1 | - | 7 | 589 | 9021497 | ||
CR749227.1 | - | 7 | 776 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
