siRNA Details
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF326987.1 | - | 3 | 475 | ||
AK304503.1 | - | 3 | 414 | |||
AK314497.1 | - | 4 | 495 | |||
AK316152.1 | - | 3 | 429 | |||
AX775917.1 | - | 4 | 527 | |||
AX775921.1 | - | 4 | 527 | |||
AY288928.1 | - | 4 | 487 | |||
BC041575.1 | - | 3 | 415 | |||
BC120996.1 | - | 4 | 505 | 40116513 | ||
DC319920.1 | - | 4 | 527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
