siRNA Details
Gene Symbol: RP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK304443.1 | - | 19 | 2303 | ||
AK316511.1 | - | 19 | 2737 |
Gene Symbol: LOC107984125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014157.1 | XP_016869646.1 | 12 | 1850 | ||
XM_017014158.1 | XP_016869647.1 | 7 | 1186 |
