siRNA Details
Gene Symbol: LOC389906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034031.1 | 2, 3 | 1449 | |||
GenBank mRNA | BC038894.1 | - | 1, 2 | 116 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC389906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034031.1 | 2, 3 | 1449 | |||
GenBank mRNA | BC038894.1 | - | 1, 2 | 116 |