siRNA Details
Gene Symbol: LOC389906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034031.1 | 5 | 1654 | |||
GenBank mRNA | BC017231.2 | - | 4 | 712 | ||
BC031508.2 | - | 3 | 215 | |||
BC035462.2 | - | 3 | 215 | |||
BC038894.1 | - | 4 | 321 |
Gene Symbol: LOC101930105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DR760352.1 | - | 2 | 324 |
Gene Symbol: LOC729162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000470.1 | - | 3 | 390 |
