siRNA Details
Gene Symbol: IMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK001830.1 | - | 1 | 786 | ||
AK057531.1 | - | 2 | 1683 | |||
AK223121.1 | - | 1 | 795 | |||
AL136913.1 | - | 1 | 783 | |||
AY050497.1 | - | 1 | 781 | |||
BC006487.1 | - | 1 | 782 | 3546883 | ||
BC011745.2 | - | 1 | 765 | 3344519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
