siRNA Details
Gene Symbol: EXOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018303.5 | NP_060773.3 | 22 | 2391 | ||
NR_073064.1 | 24 | 2553 | ||||
XM_017011018.1 | XP_016866507.1 | 23 | 2437 | |||
XM_017011019.1 | XP_016866508.1 | 23 | 2599 | |||
XM_017011020.1 | XP_016866509.1 | 23 | 2459 | |||
XM_017011021.1 | XP_016866510.1 | 22 | 2695 | |||
XM_017011022.1 | XP_016866511.1 | 22 | 2391 | |||
XM_017011023.1 | XP_016866512.1 | 22 | 2391 | |||
XM_017011024.1 | XP_016866513.1 | 23 | 2437 | |||
XM_017011025.1 | XP_016866514.1 | 22 | 2391 | |||
XM_017011026.1 | XP_016866515.1 | 22 | 2391 | |||
GenBank mRNA | AJ414403.1 | - | 22 | 2315 | ||
AK001888.1 | - | 18 | 1806 | |||
AK056564.1 | - | 22 | 2359 | |||
AK302941.1 | - | 12 | 1132 | |||
AK314628.1 | - | 22 | 2391 | |||
AL833209.1 | - | 20 | 1947 | |||
AL833213.1 | - | 22 | 2337 | |||
BC016918.1 | - | 19 | 1871 | |||
BC080542.1 | - | 24 | 2523 | 5479496 |
