siRNA Details
Gene Symbol: GLOD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080489.2 | NP_001073958.2 | 2, 3 | 235 | ||
XM_011543911.2 | XP_011542213.1 | 3, 4 | 338 | |||
XM_011543912.2 | XP_011542214.1 | 3, 4 | 482 | |||
GenBank mRNA | DY655159.1 | - | 1, 2 | 76 |
