siRNA Details
Gene Symbol: ZNF839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267827.1 | NP_001254756.1 | 8 | 2662 | ||
NM_001267828.1 | NP_001254757.1 | 8 | 2593 | |||
NM_018335.4 | NP_060805.3 | 8 | 2675 | |||
XM_006720203.3 | XP_006720266.2 | 8 | 2559 | |||
XM_011536945.1 | XP_011535247.1 | 9 | 2777 | |||
XM_011536946.2 | XP_011535248.1 | 8 | 2627 | |||
XM_011536948.2 | XP_011535250.1 | 8 | 2403 | |||
XM_017021447.1 | XP_016876936.1 | 7 | 2525 | |||
XM_017021449.1 | XP_016876938.1 | 7 | 1941 | |||
GenBank mRNA | AF155112.1 | - | 6 | 1303 | ||
AK001994.1 | - | 4 | 1091 | |||
AK093342.1 | - | 8 | 2662 | |||
AK125678.1 | - | 4 | 1811 | |||
AK127821.1 | - | 4 | 1909 | |||
AK289632.1 | - | 8 | 2593 | |||
BC073137.1 | - | 6 | 1458 |
