siRNA Details
Gene Symbol: CFAP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164496.1 | NP_001157968.1 | 35 | 6099 | ||
XM_005247617.3 | XP_005247674.1 | 18 | 4376 | |||
XM_006713696.2 | XP_006713759.1 | 36 | 6689 | |||
XM_011512977.2 | XP_011511279.1 | 34 | 6317 | |||
XM_011512978.2 | XP_011511280.1 | 31 | 5586 | |||
XM_011512979.2 | XP_011511281.1 | 30 | 5330 | |||
XM_017006844.1 | XP_016862333.1 | 35 | 6533 | |||
XM_017006845.1 | XP_016862334.1 | 31 | 5807 | |||
XM_017006846.1 | XP_016862335.1 | 30 | 6484 | |||
XM_017006847.1 | XP_016862336.1 | 31 | 5527 | |||
GenBank mRNA | AK002004.1 | - | 10 | 1981 | ||
AL133606.1 | - | 17 | 3441 | |||
BC016799.1 | - | 1 | 279 | |||
CR936646.1 | - | 4 | 1123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
