siRNA Details
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174098.1 | NP_001167569.1 | 4, 5 | 662 | ||
NM_018344.5 | NP_060814.4 | 4, 5 | 662 | |||
NR_033413.1 | 3, 4 | 579 | ||||
XM_006717910.3 | XP_006717973.1 | 4, 5 | 1054 | |||
XM_017016377.1 | XP_016871866.1 | 3, 4 | 876 | |||
GenBank mRNA | AF326987.1 | - | 3, 4 | 610 | ||
AK002022.1 | - | 3, 4 | 413 | |||
AK304503.1 | - | 3, 4 | 549 | |||
AK314497.1 | - | 4, 5 | 630 | |||
AK316152.1 | - | 3, 4 | 564 | |||
AX775915.1 | - | 4, 5 | 662 | |||
AX775917.1 | - | 4, 5 | 662 | |||
AX775919.1 | - | 4, 5 | 662 | |||
AX775921.1 | - | 4, 5 | 662 | |||
AY288928.1 | - | 4, 5 | 622 | |||
BC000223.1 | - | 1, 2 | 37 | |||
BC041575.1 | - | 3, 4 | 550 | |||
BC120996.1 | - | 4, 5 | 640 | 40116513 | ||
BC120997.1 | - | 4, 5 | 640 | 40116514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
