siRNA Details
Gene Symbol: LOC100996842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322800.1 | NP_001309729.1 | 1 | 602 | ||
NR_136403.1 | 1 | 602 | ||||
XM_006721623.3 | XP_006721686.1 | 1 | 724 | |||
GenBank mRNA | DT217613.1 | - | 1 | 602 |
