siRNA Details
Gene Symbol: LINC01344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128015.1 | - | 6 | 640 |
Gene Symbol: ZNF648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000302.1 | XP_016855791.1 | 2 | 1510 |
