siRNA Details
Gene Symbol: NEMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142645.1 | NP_001136117.1 | 3 | 431 | ||
NR_136298.1 | 3 | 431 | ||||
XM_005246212.3 | XP_005246269.1 | 3 | 585 | |||
XM_011510458.2 | XP_011508760.1 | 3 | 411 | |||
XM_011510459.2 | XP_011508761.1 | 3 | 616 | |||
XM_017003098.1 | XP_016858587.1 | 3 | 418 | |||
XM_017003099.1 | XP_016858588.1 | 3 | 418 | |||
XM_017003100.1 | XP_016858589.1 | 3 | 418 | |||
GenBank mRNA | AK296987.1 | - | 3 | 379 | ||
AK302425.1 | - | 3 | 556 |
