siRNA Details
Gene Symbol: CASC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015410.1 | 9 | 1320 | |||
GenBank mRNA | AK022865.1 | - | 8 | 946 | ||
AK026189.1 | - | 9 | 1320 | |||
BC044235.1 | - | 7 | 990 | |||
KP981381.1 | - | 6 | 834 |
