siRNA Details
Gene Symbol: LOC401320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038889.1 | 5 | 898 | |||
GenBank mRNA | AK096056.1 | - | 5 | 878 | ||
AK097240.1 | - | 2 | 1090 | |||
BC016976.1 | - | 4 | 709 | |||
BC041636.1 | - | 2 | 279 | |||
BX648714.1 | - | 3 | 479 |
