siRNA Details
Gene Symbol: GLYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308096.1 | NP_001295025.1 | 16 | 2963 | ||
NM_001324098.1 | NP_001311027.1 | 15 | 3016 | |||
NM_032569.3 | NP_115958.2 | 16 | 2981 | |||
NR_136695.1 | 18 | 2893 | ||||
NR_136696.1 | 17 | 2676 | ||||
NR_136697.1 | 17 | 2694 | ||||
NR_136698.1 | 16 | 2625 | ||||
NR_136699.1 | 16 | 2643 | ||||
NR_136700.1 | 15 | 3133 | ||||
XM_005255639.4 | XP_005255696.2 | 14 | 2731 | |||
XM_011522716.2 | XP_011521018.1 | 16 | 3109 | |||
XM_011522717.1 | XP_011521019.1 | 17 | 3166 | |||
XM_017023786.1 | XP_016879275.1 | 16 | 3059 | |||
XM_017023787.1 | XP_016879276.1 | 15 | 2993 | |||
XM_017023788.1 | XP_016879277.1 | 17 | 3148 | |||
XM_017023789.1 | XP_016879278.1 | 16 | 3115 | |||
GenBank mRNA | AF217521.1 | - | 1 | 825 | ||
AF244907.1 | - | 16 | 2962 | |||
BC032855.1 | - | 16 | 2974 | 5265561 | ||
BC047223.1 | - | 16 | 2962 | 5263991 | ||
BC048291.1 | - | 1 | 795 | |||
BC064940.1 | - | 18 | 2793 | 5787345 | ||
CR933687.1 | - | 3 | 1262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
