siRNA Details
Gene Symbol: LINC00963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038955.1 | 1 | 557 | |||
GenBank mRNA | AK024177.1 | - | 1 | 110 | ||
AK092686.1 | - | 1 | 15 | |||
BC044229.1 | - | 1 | 117 | |||
DR006520.1 | - | 1 | 563 |
