siRNA Details
Gene Symbol: LINC00963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038955.1 | 1 | 416 | |||
GenBank mRNA | DR006520.1 | - | 1 | 422 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038955.1 | 1 | 416 | |||
GenBank mRNA | DR006520.1 | - | 1 | 422 |