siRNA Details
Gene Symbol: LARP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286679.1 | NP_001273608.1 | 2 | 279 | ||
NM_018357.3 | NP_060827.2 | 2 | 544 | |||
GenBank mRNA | AF443828.1 | - | 2 | 437 | ||
AF443829.1 | - | 2 | 437 | |||
AK002058.1 | - | 2 | 438 | |||
AK130092.1 | - | 3 | 338 | |||
AL833876.1 | - | 2 | 398 | |||
BC006082.1 | - | 2 | 391 | 2961587 | ||
BC009446.1 | - | 2 | 391 | 3529516 | ||
BC014018.2 | - | 2 | 395 | 4127153 | ||
BI913258.1 | - | 2 | 452 | |||
DA715263.1 | - | 2 | 279 | |||
DN992782.1 | - | 2 | 549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
