siRNA Details
Gene Symbol: LRIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018372.3 | NP_060842.3 | 2 | 1765 | ||
XM_005271029.3 | XP_005271086.1 | 2 | 2024 | |||
XM_011541768.1 | XP_011540070.1 | 2 | 1765 | |||
XM_017001768.1 | XP_016857257.1 | 2 | 1765 | |||
XM_017001769.1 | XP_016857258.1 | 2 | 2024 | |||
GenBank mRNA | AK002131.1 | - | 1 | 706 | ||
AL834466.1 | - | 1 | 683 | |||
AY190122.1 | - | 2 | 1755 | |||
BC008115.1 | - | 1 | 907 | 2900378 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
