siRNA Details
Gene Symbol: LINC00865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038382.1 | 4 | 2421 | |||
GenBank mRNA | AL137595.1 | - | 3 | 2023 | ||
BC033541.1 | - | 4 | 2404 | |||
BC035398.1 | - | 4 | 2404 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038382.1 | 4 | 2421 | |||
GenBank mRNA | AL137595.1 | - | 3 | 2023 | ||
BC033541.1 | - | 4 | 2404 | |||
BC035398.1 | - | 4 | 2404 |