siRNA Details
Gene Symbol: LOC100288637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038253.1 | 6 | 636 | |||
NR_038254.1 | 5 | 536 | ||||
NR_038255.1 | 4 | 441 | ||||
GenBank mRNA | BC112921.1 | - | 4 | 313 | ||
BX954662.1 | - | 4 | 419 | |||
BX954797.1 | - | 5 | 537 |
