siRNA Details
Gene Symbol: ST6GALNAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018414.4 | NP_060884.1 | 1, 2 | 339 | ||
NR_110309.1 | 1, 2 | 339 | ||||
XM_011524995.2 | XP_011523297.1 | 1, 2 | 259 | |||
XM_011524996.2 | XP_011523298.1 | 1, 2 | 259 | |||
XM_017024842.1 | XP_016880331.1 | 1, 2 | 259 | |||
XM_017024843.1 | XP_016880332.1 | 1, 2 | 259 | |||
GenBank mRNA | AK310906.1 | - | 1, 2 | 275 | ||
AY096001.1 | - | 1, 2 | 139 | |||
AY358918.1 | - | 1, 2 | 284 | |||
BC022462.1 | - | 1, 2 | 194 | 4816713 | ||
DA594489.1 | - | 1, 2 | 339 | |||
Y11339.2 | - | 1, 2 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
