siRNA Details
Gene Symbol: LINC01578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037600.1 | 3 | 446 | |||
NR_037601.1 | 4 | 640 | ||||
NR_037602.1 | 3 | 350 | ||||
GenBank mRNA | AA083786.1 | - | 4 | 320 | ||
BQ188346.1 | - | 3 | 203 | |||
BQ890654.1 | - | 3 | 350 | |||
BU848955.1 | - | 3 | 438 |
