siRNA Details
Gene Symbol: SHISA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145205.1 | NP_001138677.2 | 1, 2 | 1016 | ||
GenBank mRNA | AK309038.1 | - | 1, 2 | 357 | ||
CR627300.1 | - | 1, 2 | 212 | |||
CR749545.1 | - | 1, 2 | 212 |
