siRNA Details
Gene Symbol: LOC100134317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029389.1 | 2 | 507 |
Gene Symbol: LOC284412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029390.1 | 2 | 2254 | |||
GenBank mRNA | AF289566.1 | - | 1 | 2221 | ||
BC045185.1 | - | 2 | 2261 |
