siRNA Details
Gene Symbol: LINC01136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034150.1 | 3 | 1199 | |||
NR_034151.1 | 4 | 1385 | ||||
GenBank mRNA | AK024293.1 | - | 3 | 1183 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034150.1 | 3 | 1199 | |||
NR_034151.1 | 4 | 1385 | ||||
GenBank mRNA | AK024293.1 | - | 3 | 1183 |