siRNA Details
Gene Symbol: RNLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018363.3 | NP_060833.1 | 7 | 2008 | ||
XM_005269950.3 | XP_005270007.1 | 5 | 2294 | |||
GenBank mRNA | AK002080.1 | - | 7 | 1935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
