siRNA Details
Gene Symbol: LOC100996693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286811.1 | NP_001273740.1 | 3 | 451 | ||
XM_011510469.2 | XP_011508771.1 | 4 | 2932 | |||
GenBank mRNA | DA086253.1 | - | 2 | 257 |
