siRNA Details
Gene Symbol: THEGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534357.1 | XP_011532659.1 | 9 | 1446 | ||
XM_011534361.2 | XP_011532663.1 | 9 | 1446 | |||
XM_011534363.2 | XP_011532665.1 | 9 | 1446 | |||
XM_011534364.1 | XP_011532666.1 | 10 | 1481 |
