siRNA Details
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018396.2 | NP_060866.2 | 9 | 1468 | ||
GenBank mRNA | AK002212.1 | - | 8 | 1342 | ||
AK299960.1 | - | 9 | 1432 | |||
BC064929.1 | - | 9 | 1428 | |||
BC107586.1 | - | 8 | 1352 | 6494734 |
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181725.3 | NP_859076.3 | 9 | 1448 | ||
XM_017024572.1 | XP_016880061.1 | 8 | 1329 | |||
GenBank mRNA | AK000991.1 | - | 4 | 695 | ||
AK022822.1 | - | 9 | 1425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
